ContactUsLinkComponent
Silica gel 3-6 mm, non toxic for drying
Silica gel 3-6 mm, non toxic for drying
Certificates
undefined
Silica gel 3-6 mm, non toxic for drying
Certificates
Specification Test Results
  • Appearance:
    Colorless to white Irregular granules
  • Particle size:
    3.0 to 6.0 mm
  • Residual water:
    ≤2.0 % (140°C, 4 h)
  • Water adsorption:
    ≥20.0 % (40% rH)