Order Entry
Czech Republic
ContactUsLinkComponent
Tin(II) chloride, anhydrous ≥99%, GPR RECTAPUR®
Tin(II) chloride, anhydrous ≥99%, GPR RECTAPUR®
Catalog # 23746.296
Supplier:  VWR Chemicals
CAS Number:  
undefined
Tin(II) chloride, anhydrous ≥99%, GPR RECTAPUR®
Catalog # 23746.296
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • SO4 (Sulphate):
    Max. 0.02 %
  • Cd (Cadmium):
    Max. 40 ppm
  • Cu (Copper):
    Max. 40 ppm
  • Fe (Iron):
    Max. 100 ppm
  • Pb (Lead):
    Max. 75 ppm
  • Zn (Zinc):
    Max. 40 ppm

Specification Test Results

About this item

 :
 :189.62 g/mol
Merck Index:13,08861
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:3.95 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-868-0
UN:3260
ADR:8,III
 :GHS07
 :GHS05
CAS Number:7772-99-8
Formula:SnCl₂
MDL Number:MFCD00011241