Order Entry
Czech Republic
ContactUsLinkComponent
Orthoboric acid, Suprapur® for trace analysis, Supelco®
Health
Orthoboric acid, Suprapur® for trace analysis, Supelco®
Catalog # 1.00765.0050
Supplier:  Merck
CAS Number:  
undefined
Health
Orthoboric acid, Suprapur® for trace analysis, Supelco®
Catalog # 1.00765.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    46.5 g/L (20 °C)
  • Melting Point:
    171 °C (decomposition)
  • Molar Mass:
    61.83 g/mol
  • Bulk Density:
    400 - 600 kg/m³
  • Vapour Pressure:
    2.7 hPa (20 °C)
  • Density:
    1.44 g/cm³ (20 °C)
  • pH:
    3.7 (46.5 g/L, H₂O, 20 °C)

Specification Test Results

About this item

 :
Merck Index:13,01326
Boiling Pt:185 °C (1013 hPa)
Melting Pt:160 °C
Density:1.44 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ100
 :61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
REACH:01-2119486683-25
 :GHS08
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337