

Kyselina boritá ≥99.5% (by acidimetry), granulated, Ultrapure Bioreagent for molecular biology, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:10043-35-3
:
Formula:B(OH)₃
MDL Number:MFCD00011337
Merck Index:13,01326
Boiling Pt:185 °C (1013 hPa)
Melting Pt:160 °C
Density:1,44 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:233-139-2
REACH:01-2119486683-25
:GHS07
:GHS08
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.5 %
- Assay (H₃BO₃) (by acidimetry):≥99.5 %
- RNase Activity:Passes Test
- DNase Activity (None Detected):Passes Test
- Protease Activity:Passes Test
- Chloride (Cl):≤0.001 %
- Phosphate (PO₄):≤0.001 %
- Sulfate (SO₄):≤0.005 %
- Calcium (Ca):≤0.005 %
- Heavy Metals (as Pb):≤0.001 %
- Insoluble in Methanol:≤0.005 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Trace Impurities - Arsenic (As):≤1.000 ppm
- Trace Impurities - Magnesium (Mg):≤5.000 ppm
Specification Test Results
Frequently Bought Together