
Diboron trioxide, Sigma-Aldrich®
Catalog # SIAL11615-1KG-R
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:69.62 g/mol
Merck Index:13,01327
Boiling Pt:1860 °C (1013 hPa)
Melting Pt:450 °C
Density:1.81…1.86 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:215-125-8
REACH:01-2119486655-24
:GHS08
CAS Number:1303-86-2
Formula:B₂O₃
MDL Number:MFCD00011315