Order Entry
Czech Republic
ContactUsLinkComponent
Diboron trioxide ≥99.9% (B basis)
Health
Diboron trioxide ≥99.9% (B basis)
Catalog # ABCRAB119517.0050
Supplier:  abcr
CAS Number:  
Health
Diboron trioxide ≥99.9% (B basis)
Catalog # ABCRAB119517.0050
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

About this item

 :
 :69.62 g/mol
Merck Index:13,01327
Boiling Pt:~1860 °C
Melting Pt:450 °C
Density:2.46
Storage Temperature:Ambient
EINECS:215-125-8
REACH:01-2119486655-24
 :GHS08
CAS Number:1303-86-2
Formula:B₂O₃
MDL Number:MFCD00011315