syn-Pyridine-2-carboxaldoxime, Sigma-Aldrich®
Catalog # SIALP58200-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:122.13 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:1193-96-0
Formula:C₆H₆N₂O
MDL Number:MFCD00167166