3-Methyl-1-phenyl-2-pyrazolin-5-one, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:89-25-8
:
Formula:C₁₀H₁₀N₂O
MDL Number:MFCD00003138
:174.2 g/mol
:M-Clarity™ quality level = MQ200
EINECS:201-891-0
Frequently Bought Together