N,N-Dimethylaminophosphoryl dichloride, Sigma-Aldrich®
Katalogové číslo SIAL79625-10ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
ADR:8,II
:161.95 g/mol
EINECS:211-641-2
UN:3265
CAS Number:677-43-0
Formula:C₂H₆Cl₂NOP
MDL Number:MFCD00013896