Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1'-Dibromoferrocene
1,1'-Dibromoferrocene, Sigma-Aldrich®
1,1'-Dibromoferrocene, Sigma-Aldrich®
SIAL710393-1G
:
Merck
:
1,1'-Dibromoferrocene, Sigma-Aldrich®
SIAL710393-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
335.76 g/mol
:
1293-65-8
:
C₁₀Br₂Fe
:
MFCD01075755