Indium(III) chloride, anhydrous 98+%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 10025-82-8 Molecular Formula: InCl3 Formula Weight: 221.18 Odour: Odourless Melting Point: 586° Boiling Point: 800° Merck Reference: 14,4958 UN No.: 3260 MDL No.: MFCD00011058
Synonyms: Indium Trichloride Chloride, Indium trichloride
- CAS Number:10025-82-8
- Formula:Cl₃In
- MDL Number:MFCD00011058
- Molecular Weight:221.18 g/mol
- Merck Index:14,4958
- Boiling Pt:800 °C
- Melting Pt:586 °C
- Density:3.460
- Storage Temperature:Ambient
- EINECS:233-043-0
- UN:3260
- ADR:8,II
2 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 10025-82-8 Molecular Formula: InCl3 Formula Weight: 221.18 Odour: Odourless Melting Point: 586° Boiling Point: 800° Merck Reference: 14,4958 UN No.: 3260 MDL No.: MFCD00011058
Synonyms: Indium Trichloride Chloride, Indium trichloride
- CAS Number:10025-82-8
- Formula:Cl₃In
- MDL Number:MFCD00011058
- Molecular Weight:221.18 g/mol
- Merck Index:14,4958
- Boiling Pt:800 °C
- Melting Pt:586 °C
- Density:3.460
- Storage Temperature:Ambient
- EINECS:233-043-0
- UN:3260
- ADR:8,II
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



