Indium(III) chloride, anhydrous ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Indium Trichloride Chloride, Indium trichloride
CAS Number:10025-82-8
:
Formula:Cl₃In
MDL Number:MFCD00011058
Boiling Pt:800 °C
Melting Pt:586
Density:3.460
Molecular Weight:221.18 g/mol
Storage Temperature:Ambient
EINECS:233-043-0
ADR:8 II
Merck Index:14,4958
UN:3260
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together