1,3-Butanediol diacrylate, Sigma-Aldrich®
Katalogové číslo SIAL411752-25ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:198.22 g/mol
:M-Clarity™ quality level = MQ100
EINECS:243-105-9
CAS Number:19485-03-1
Formula:C₁₀H₁₄O₄