Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Butanediol dimethacrylate
1,3-Butanediol dimethacrylate, Sigma-Aldrich®
1,3-Butanediol dimethacrylate, Sigma-Aldrich®
1,3-Butanediol dimethacrylate, Sigma-Aldrich®
SIAL408999-250ML
:
Merck
:
1,3-Butanediol dimethacrylate, Sigma-Aldrich®
SIAL408999-250ML
:
Merck
:
Pack type:
Glass bottle
Pk:
250 mL
Seq:
2590
:
:
226.27 g/mol
:
M-Clarity™ quality level = MQ100
:
214-711-0
:
1189-08-8
:
C₁₂H₁₈O₄
:
MFCD00014930