2-(Hydroxymethyl)-1,3-propanediol, Sigma-Aldrich®
Catalog # SIAL393657-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:106.12 g/mol
:M-Clarity™ quality level = MQ100
EINECS:225-187-8
CAS Number:4704-94-3
Formula:C₄H₁₀O₃
MDL Number:MFCD00192217