N,N'-Dimethyl-1,3-propanediamine, Sigma-Aldrich®
Catalog # SIAL308110-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:102.18 g/mol
:M-Clarity™ quality level = MQ200
EINECS:203-859-1
CAS Number:111-33-1
Formula:C₅H₁₄N₂
MDL Number:MFCD00008292