Di-tert-butyl-3-oxoglutarate, Sigma-Aldrich®
Catalog # SIAL270814-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:258.31 g/mol
:M-Clarity™ quality level = MQ200
EINECS:248-775-6
CAS Number:28009-80-5
Formula:C₁₃H₂₂O₅