Order Entry
Czech Republic
ContactUsLinkComponent
Palladium 5% on barium carbonate, reduced, Sigma-Aldrich®
Palladium 5% on barium carbonate, reduced, Sigma-Aldrich®
Catalog # SIAL237523-10G
Supplier:  Merck
CAS Number:  
Palladium 5% on barium carbonate, reduced, Sigma-Aldrich®
Catalog # SIAL237523-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :106.420 g/mol
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
EINECS:231-115-6
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD00011167