N,N-Dimethylthioformamide, Sigma-Aldrich®
Catalog # SIAL163643-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:89.16 g/mol
:M-Clarity™ quality level = MQ100
EINECS:212-060-7
CAS Number:758-16-7
Formula:C₃H₇NS
MDL Number:MFCD00004943