
(±)-2,3-Epoxypropyl methacrylate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-91-2
:
Formula:C₇H₁₀O₃
MDL Number:MFCD00005137
:142.15 g/mol
ADR:6.1,III
:M-Clarity™ quality level = MQ200
EINECS:203-441-9
UN:2810
:GHS07
Frequently Bought Together