Order Entry
Czech Republic
ContactUsLinkComponent
Gallium(II) chloride, ultra dry ≥99.999% (metals basis)
Corrosive
Gallium(II) chloride, ultra dry ≥99.999% (metals basis)
Catalog # 35696.03
CAS Number:  
Corrosive
Gallium(II) chloride, ultra dry ≥99.999% (metals basis)
Catalog # 35696.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :281.26 g/mol
Boiling Pt:535 °C
Melting Pt:164 °C
Storage Temperature:Ambient
ADR:8,II
 :ampouled under argon
EINECS:246-338-4
UN:3260
 :GHS05
CAS Number:13498-12-9
 :24597-12-4
Formula:Ga₂Cl₄
MDL Number:MFCD00151526