Order Entry
Czech Republic
ContactUsLinkComponent
Isonicotinaldehyde oxime
Irritant
Isonicotinaldehyde oxime
Catalog # 8.20714.0050
Supplier:  Merck
CAS Number:  
Irritant
Isonicotinaldehyde oxime
Catalog # 8.20714.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    10 g/l (20 °C)
  • Melting Point:
    130 - 133 °C
  • Molar Mass:
    122.12 g/mol
  • Bulk Density:
    610 kg/m3
  • Vapor Pressure:
     - 0.01 hPa (50 °C)
  • Flash Point:
    110 °C DIN 51758
  • pH:
    6.3 (10 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :122.13 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:211-796-6
 :GHS07
Storage Temperature:Ambient
CAS Number:696-54-8
Formula:C₆H₆N₂O
MDL Number:MFCD00006446