Order Entry
Czech Republic
ContactUsLinkComponent
Triethylene glycol monomethyl ether, Sigma-Aldrich®
Triethylene glycol monomethyl ether, Sigma-Aldrich®
Catalog # 8.14587.1000
Supplier:  Merck
CAS Number:  
Triethylene glycol monomethyl ether, Sigma-Aldrich®
Catalog # 8.14587.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Molar Mass:
    164.2 g/mol
  • Boiling Point:
    245 - 255 °C (1013 hPa)
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Flash Point:
    133 °C
  • Density:
    1.04 g/cm3 (20 °C)
  • pH:
    5 - 7 (H2O, 20 °C)
  • Ignition Point:
    210 °C

Specification Test Results

About this item

Synonyms: mPEG3-Alcohol, 2-(2-(2-Methoxyethoxy)ethoxy)ethanol, Methyl triglycol, m-PEG3-OH
 :
 :164.2 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:203-962-1
CAS Number:112-35-6
Formula:C₇H₁₆O₄
MDL Number:MFCD00002875