
Aluminium chloride, anhydrous ≥99%, GPR RECTAPUR®
Catalog # 21031.293
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:Min. 99 %
- Heavy metals (as Pb):Max. 20 ppm
- Insolubility in water:Max. 100 ppm
- Not precipitated by NH4OH (as SO4):Max. 0.1 %
- SO4 (Sulphate):Max. 100 ppm
- Fe (Iron):Max. 100 ppm
Specification Test Results
About this item
:
:133.34 g/mol
Merck Index:13,00335
Boiling Pt:180 °C (1013 hPa)
Melting Pt:180…181 °C
Density:1.31 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-208-1
UN:1726
ADR:8,II
:GHS05
CAS Number:7446-70-0
Formula:AlCl₃
MDL Number:MFCD00003422