Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
 
1,3-Dibromo-5,5-dimethylhydantoin, Sigma-Aldrich®
  8.03600.0250
 :  Merck
 :  
 
 
 
 
1,3-Dibromo-5,5-dimethylhydantoin, Sigma-Aldrich®
  8.03600.0250
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

 

 

  • Solubility:
    1 g/l (20 °C)
  • Melting Point:
    186.0 - 192.0 °C
  • Molar Mass:
    285.92 g/mol
  • Flash Point:
    155.0 °C
  • Density:
    1.00 g/cm3 (25 °C)
  • Ignition Point:
    550 °C

 

 

 :
 :8 (5.1),II
 :M-Clarity™ quality level = MQ200
 :285.92 g/mol
 :201-030-9
 :3084
 :GHS03
 :GHS06
 :GHS05
 :GHS09
 :Ambient
 :77-48-5
 :C₅H₆Br₂N₂O₂
 :MFCD00003189