


N,N-Dimethyl-1,3-diaminopropane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:109-55-7
:
Formula:C₅H₁₄N₂
MDL Number:MFCD00008216
:102.18 g/mol
ADR:8 (3),II
:M-Clarity™ quality level = MQ200
EINECS:203-680-9
UN:2734
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:102.17 g/mol
- Boiling Point:132 - 140 °C (1013 hPa)
- Vapor Pressure:17 hPa (30 °C)
- Flash Point:32 °C
- Refractive Index:1.4415 (20 °C, 589 nm)
- Explosion Limit:2.3 - 12.35 %(V)
- Density:0.82 g/cm3 (20 °C)
- pH:12.7 (100 g/l, H2O, 20 °C)
- Ignition Point:215 °C
Specification Test Results
Frequently Bought Together