
Cystamine dihydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:56-17-7
:
Formula:C₄H₁₂N₂S₂·2HCl
MDL Number:MFCD00012905
:225.20 g/mol
:M-Clarity™ quality level = MQ200
EINECS:200-260-7
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:218 - 221 °C
- Molar Mass:225.19 g/mol
- Synonyms:Cystamine dihydrochloride
- Hill Formula:C4H14Cl2N2S2
- Chemical Formula:[H2NCH2CH2SSCH2CH2NH2] * 2 HCl
Specification Test Results
Frequently Bought Together