Order Entry
Czech Republic
ContactUsLinkComponent
(±)-Allyl-2,3-epoxypropyl ether, Sigma-Aldrich®
Flammable
Health
Corrosive
Irritant
(±)-Allyl-2,3-epoxypropyl ether, Sigma-Aldrich®
Catalog # 8.00259.0250
Supplier:  Merck
CAS Number:  
Flammable
Health
Corrosive
Irritant
(±)-Allyl-2,3-epoxypropyl ether, Sigma-Aldrich®
Catalog # 8.00259.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    50 g/l (20 °C)
  • Melting Point:
    -100 °C
  • Molar Mass:
    114.15 g/mol
  • Boiling Point:
    154 °C (1013 hPa)
  • Vapor Pressure:
    2.6 hPa (20 °C)
  • Flash Point:
    45 °C
  • Refractive Index:
    1.4332 (20 °C, 589 nm)
  • Density:
    0.97 g/cm3 (20 °C)
  • Ignition Point:
    264 °C

Specification Test Results

About this item

 :
 :114.14 g/mol
ADR:3,III
 :M-Clarity™ quality level = MQ200
EINECS:203-442-4
UN:2219
 :GHS02
 :GHS08
 :GHS05
 :GHS07
Storage Temperature:Refrigerator
CAS Number:106-92-3
Formula:C₆H₁₀O₂
MDL Number:MFCD00005143