
N,N-Dimethylaminophosphoryl dichloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:677-43-0
:
Formula:C₂H₆Cl₂NOP
MDL Number:MFCD00013896
Boiling Pt:89…90 °C (20 mmHg)
Density:1.360
Flash Pt:>110 °C (230 °F)
EINECS:211-641-2
UN:3265
ADR:8,II
:161.95 g/mol
:GHS05
Related Information
Frequently Bought Together