0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Glyoxylic acid
Glyoxylic acid 50% (w/w) in aqueous solution
Glyoxylic acid 50% (w/w) in aqueous solution
:
Thermo Fisher Scientific
Glyoxylic acid 50% (w/w) in aqueous solution
:
Thermo Fisher Scientific
:
298-12-4
:
:
C₂H₂O₃
:
MFCD00006958
:
74.04 g/mol
:
111 °C
:
–93 °C
:
1.339
:
Refrigerator
:
206-058-5
:
3265
:
8,III
:
GHS05
:
GHS07
msds_ALFAB25149
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
: Merck
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
: VWR Chemicals
Acetone, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Oxalic acid dihydrate for synthesis, Sigma-Aldrich®
: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals