
1-Phenyl-3-(trifluoromethyl)-2-pyrazolin-5-one ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:321-07-3
:
Formula:C₁₀H₇F₃N₂O
MDL Number:MFCD00051655
:228.17 g/mol
Melting Pt:191…195 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together