3-Quinuclidinone hydrochloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1193-65-3
:
Formula:C₇H₁₂ClNO
MDL Number:MFCD00137391
:161.63 g/mol
Melting Pt:∼ 300 °C (dec.)
Storage Temperature:Ambient
EINECS:214-776-5
Related Information
Frequently Bought Together