
3-Methyl-1-phenyl-2-phospholene-1-oxide ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:707-61-9
:
Formula:C₁₁H₁₃OP
MDL Number:MFCD00014518
:192.2 g/mol
Boiling Pt:173…174 °C (0.7 mmHg)
Melting Pt:60…65 °C
Storage Temperature:Ambient
EINECS:211-901-5
:GHS07
Related Information
Frequently Bought Together