

N,N'-Dimethyl-1,3-propanediamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-33-1
:
Formula:C₅H₁₄N₂
MDL Number:MFCD00008292
:102.18 g/mol
Boiling Pt:145 °C
Density:0.81 g/cm³
Flash Pt:20 °C
Storage Temperature:Ambient
EINECS:203-859-1
UN:2733
ADR:3,II
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4370 to 1.4410 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together