
Palladium (5% Pd) on activated carbon, unreduced
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD00011167
:106.420 g/mol
Storage Temperature:Ambient
EINECS:231-115-6
UN:3178
ADR:4.1,III
:GHS02
Specification Test Results
- Appearance:Black Powder
- Assay:4.8 to 5.2 % (Pd) On dry basis
- Loss on drying:≤3 % (120°C)
Specification Test Results
Frequently Bought Together