
syn-Pyridine-2-carboxaldoxime 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1193-96-0
:
Formula:C₆H₆N₂O
MDL Number:MFCD00167166
:122.13 g/mol
Melting Pt:110…113 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light pink Crystalline powder
- Infrared spectrum:Conforms
- Melting point:110 to 113 °C
- Water:≤0.2 % (K.F.)
- Titration with HClO4:99.0 to 101.0 %
Specification Test Results
Frequently Bought Together