



(±)-Allyl-2,3-epoxypropyl ether 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-92-3
:
Formula:C₆H₁₀O₂
MDL Number:MFCD00005143
:114.14 g/mol
Boiling Pt:154 °C (760 mmHg)
Melting Pt:–100 °C
Density:0.962 g/cm³
Flash Pt:48 °C
EINECS:203-442-4
UN:2219
ADR:3,III
:GHS06
:GHS08
:GHS05
:GHS02
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4332 to 1.4352 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together