

Strontium nitrate, anhydrous 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10042-76-9
:
Formula:Sr(NO₃)₂
MDL Number:MFCD00011248
:211.63 g/mol
Merck Index:14,8845
Boiling Pt:645 °C (1013 hPa)
Melting Pt:570 °C
Density:2.990
Storage Temperature:Ambient
EINECS:233-131-9
UN:1507
ADR:5.1,III
:GHS03
:GHS07
Related Information
Frequently Bought Together