

Barium nitrate, EMSURE® ACS for analysis, Supelco®
Catalog # 1.01729.0500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Solubility:90 g/l (20 °C)
- Melting Point:592 °C
- Molar Mass:261.34 g/mol
- Bulk Density:1900 g/m3
- Vapor Pressure:(20 °C)
- Density:3.2 g/cm3 (20 °C)
- pH:5.2 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
About this item
:
Merck Index:13,00986
Melting Pt:∼592 °C
Density:∼3.23 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1446
ADR:5.1 (6.1),II
:M-Clarity™ quality level = MQ300
:261.34 g/mol
EINECS:233-020-5
:GHS03
:GHS06
CAS Number:10022-31-8
Formula:Ba(NO₃)₂
MDL Number:MFCD00003442