Magnesium chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7786-30-3
:
Formula:MgCl₂
MDL Number:MFCD00011106
:95.21 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:715 °C
Density:2.32 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:232-094-6
Specification Test Results
- Solubility:542 g/l (20 °C)
- Melting Point:712 °C
- Molar Mass:95.22 g/mol
- Boiling Point:1412 °C (1013 hPa)
- Density:2.32 g/cm3 (20 °C)
- pH:=7 (50 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.14733.0500_TDS_EN ProductPage_814733
Product specification
msds_814733
SDS
Frequently Bought Together