Calcium chloride ≥94%, granules, GPR RECTAPUR® 2-5 mm, purified
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10043-52-4
:
Formula:CaCl₂
MDL Number:MFCD00010903
Merck Index:14,01659
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:772 °C
Density:2.174 g/cm³ (20 °C)
:110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
REACH:01-2119494219-28
:GHS07
Specification Test Results
- Assay:Min. 94 %
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Plastic bucket for solids UN Y (ZPACK13247010) 1L Plastic bottle for solids DIN 60 UN (ZPACK13251000)
Packaging Information
spec_VWRC22328.262 spec_VWRC22328.364
Product specification
VWR® for Drying and Filtration
Product Literature
Frequently Bought Together