

Sodium metavanadate (≥38% V) ≥96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13718-26-8
:
Formula:NaO₃V
MDL Number:MFCD00011125
:121.93 g/mol
Merck Index:14,8700
Melting Pt:630 °C
Density:5.15 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:237-272-7
UN:3285
ADR:6.1,III
:GHS06
:GHS07
Frequently Bought Together