ContactUsLinkComponent
Dropping bottle with head stopper
Dropping bottle with head stopper
Catalog # MARI3610105
Supplier:  MARIENFELD
undefined
Dropping bottle with head stopper
Catalog # MARI3610105
Supplier:  MARIENFELD
Specifications
  • Capacity:
    100 ml
  • Pk:
    1
Specifications
About this item
Glass, amber.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Amber coloured to protect light sensitive substanzes
Dropping bottle with flat head stopper and round shoulder.