SGE BPX90 GC Capillary Columns
Supplier: Trajan Scientific and Medical
Certificates
About this item
The SGE BPX90 GC capillary column provides a great polar option for GC analyses. Recommended for fast separation of fragrances, aromatics, petrochemical, pesticides, PCBs, isomers of Fatty Acid Methyl Esters (FAMEs).
- Unique bonded phase
- Highly polar
- Thermally stable
- Excellent resolution for cis and trans isomers
- Able to be solvent rinsed
The names and trademarks of third party analytical instrument manufacturers are used solely to indicate the compatibility of Trajan products for use with such instruments.
Specifications
- Catalog No:
- 568-0416
- 558-0349
- 558-0351
- 558-0348
- 558-0347
- Polarity:
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Temperature limit:
- 280 °C
- 280 °C
- 280 °C
- 280 °C
- 280 °C