Order Entry
Czech Republic
ContactUsLinkComponent
Sodium sulphate, anhydrous, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Sodium sulphate, anhydrous, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
undefined
Sodium sulphate, anhydrous, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    44.55 g/l (20 °C)
  • Melting Point:
    888 °C
  • Molar Mass:
    142.04 g/mol
  • Bulk Density:
    1400 - 1600 kg/m3
  • Density:
    2.70 g/cm3 (20 °C)
  • pH:
    4 - 7 (200 g/l, H2O, 20 °C)

Specification Test Results

Frequently Bought Together