Zadání objednávky
Czech Republic
ContactUsLinkComponent
Sulfit sodný, anhydrous 98+% ACS
Sulfit sodný, anhydrous 98+% ACS
Certificates
undefined
Sulfit sodný, anhydrous 98+% ACS

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Titration Iodimetric:
    ≥98.0 %
  • Free acid:
    Passes test
  • Chloride (Cl):
    ≤200 ppm
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Titrable free base:
    ≤0.03 meq/g
  • Insoluble matter:
    ≤0.005 %

Specification Test Results

Related Information

Frequently Bought Together