Sodium dihydrogen phosphate, Suprapur® for trace analysis, Supelco®
Catalog # 1.06370.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Solubility:850 g/l (20 °C)
- Molar Mass:119.98 g/mol
- Bulk Density:940 kg/m3
- pH:4.5 (10 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
Merck Index:12,08806
Melting Pt:∼650 °C
Density:0.9996 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:119.98 g/mol
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
CAS Number:7558-80-7
Formula:NaH₂PO₄
MDL Number:MFCD00003527