
Disodium hydrogen phosphate, anhydrous ≥99.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7558-79-4
:
Formula:Na₂HPO₄
MDL Number:MFCD00003496
:141.96 g/mol
Merck Index:14,8659
Melting Pt:250 °C
Density:1.53 g/cm³ (20 °C)
EINECS:231-448-7
REACH:01-2119489797-11
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together