

Sodium hexafluorophosphate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21324-39-0
:
Formula:F₆NaP
MDL Number:MFCD00011122
Melting Pt:>200 °C
Density:2.37
Storage Temperature:Ambient
EINECS:244-333-1
UN:3260
ADR:8,II
:167.95 g/mol
:GHS05
:GHS07
Related Information
Frequently Bought Together