Sodium hydroxide, BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 0404.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:min. 97 %
- Carbonate (as Na2CO3):max. 1 %
- Aluminium (Al):max. 0.002 %
- Chloride (Cl):max. 0.01 %
- Iron (Fe):max. 0.002 %
- Heavy Metals (as Pb):max. 0.002 %
- Sulfate (SO4):max. 0.01 %
Specification Test Results
About this item
Synonyms: Caustic soda
:
:40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
REACH:01-2119457892-27
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548